Learn More
Sino Biological Recombinant Human PD-1 Protein (ECD, Asp26-Pro39 deletion, His Tag), HPLC-verified
Supplier:  Sino Biological 10377H08H11
Encompass_Preferred
A DNA sequence encoding the Human PDCD1 (NP_005009.2) (Met1-Gln167,Asp26-Pro39 deletion) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3346