Learn More
Sino Biological Recombinant Human PD-L2/B7-DC Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10292H08HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Pro219) of human PD-L2 (NP_079515.2) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5390