Learn More
Sino Biological Recombinant Human PD-L2/B7-DC Protein (ECD, mFc Tag)
Supplier:  Sino Biological 10292H38H500
Encompass_Preferred
A DNA sequence encoding the human PDCD1LG2 (NP_079515.2) (Met1-Phe219) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-255-5387