Learn More
BIOHIPPO Human PDCD6IP Microsample
SDP
Catalog No. 5000314937
Encompass
Human PDCD6IP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PDCD6IP in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 117 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PDCD6IP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PDCD6IP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PDCD6IP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14937 Supplier BIOHIPPO Supplier No. BHE15211609
Human PDCD6IP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PDCD6IP in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 117 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PDCD6IP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PDCD6IP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PDCD6IP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color