Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PDDC1 (aa 119-220) Control Fragment Recombinant Protein

Catalog No. RP89869
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP89869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP89869 Supplier Invitrogen™ Supplier No. RP89869
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52613. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The PDDC1 (Parkinson Disease 7 Domain Containing 1) gene encodes a protein that is part of the DJ-1 protein family, known for its role in protecting cells against oxidative stress and neurodegenerative conditions. PDDC1 has been implicated in various cellular processes such as transcription regulation, response to oxidative damage, and cellular homeostasis. It is located on human chromosome 12 and is expressed in several tissues, including the brain. Although its exact function remains to be fully elucidated, PDDC1 shares homology with Parkinson's disease-associated protein DJ-1, suggesting a potential role in neuroprotection. Research indicates that PDDC1 might be involved in neural development and survival, and its dysregulation could contribute to diseases involving oxidative stress and neurodegeneration.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NB37
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 347862
Name Human PDDC1 (aa 119-220) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias GATD1; Glutamine amidotransferase-like class 1 domain-containing protein 1; Parkinson disease 7 domain containing 1; Parkinson disease 7 domain-containing protein 1; PDDC1; RGD1304836
Common Name PDDC1
Gene Symbol GATD1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KPICAVGHGVAALCCATNEDRSWVFDSYSLTGPSVCELVRAPGFARLPLVVEDFVKDSGACFSASEPDAVHVVLDRHLVTGQNASSTVPAVQNLLFLCGSRK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.