Learn More
Sino Biological Anti-PDE1C Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13605T08100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PDE1C (rh PDE1C; Catalog#13605-H20B; Q8TAE4; Met1-Glu634). PDE1C specific IgG was purified by Human PDE1C affinity chromatography.
Catalog No. 50-240-8966