Learn More
Sino Biological Anti-PDE2A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13031RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PDE2A (rh PDE2A; Catalog#13031-H08B; O00408-1; Glu215-His900). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1660