Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PDE4D (aa 736-790) Control Fragment Recombinant Protein

Catalog No. RP96826
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96826 Supplier Invitrogen™ Supplier No. RP96826
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111224. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes one of four mammalian counterparts to the fruit fly 'dunce' gene. The encoded protein has 3',5'-cyclic-AMP phosphodiesterase activity and degrades cAMP, which acts as a signal transduction molecule in multiple cell types. This gene uses different promoters to generate multiple alternatively spliced transcript variants that encode functional proteins.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q08499
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 5144
Name Human PDE4D (aa 736-790) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 9630011N22Rik; ACRDYS2; cAMP-specific 3',5'-cyclic phosphodiesterase 4D; cAMP-specific phosphodiesterase 4D; cAMP-specific phosphodiesterase PDE4D; cAMP-specific phosphodiesterase PDE4D6; cAMP-specific phosphodiesterase splice variant PDE4D8; cyclic AMP specific phosphodiesterase PDE4D5A; DKFZp686M11213; Dpde3; dunce; FLJ97311; HGNC:8783; HS PDE4DN2; HSPDE4D; PDE43; Pde4d; PDE4D3; PDE4D7; PDE4DN2; phosphodiesterase 4D; Phosphodiesterase 4D cAMP-specific (dunce; phosphodiesterase 4D, cAMP specific; phosphodiesterase 4D, cAMP-specific; Phosphodiesterase 4D, cAMP-specific (dunce; phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog, Drosophila); STRK1; testicular tissue protein Li 136
Common Name PDE4D
Gene Symbol Pde4d
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FELTLEEDGESDTEKDSGSQVEEDTSCSDSKTLCTQDSESTEIPLDEQVEEEAVG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.