Learn More
Sino Biological Recombinant Human PDGF-BB Protein (MALS-verified), MALS-verified
Catalog No. 502992882
Encompass_Preferred
A DNA sequence encoding the Human PDGFBB (NP_002599.1) (Ser82-Thr190) was expressed with a N-terminal Met.
Catalog No. 50-299-2882 Supplier Sino Biological Supplier No. 10572HNAE100
A DNA sequence encoding the Human PDGFBB (NP_002599.1) (Ser82-Thr190) was expressed with a N-terminal Met.