Learn More
Sino Biological Anti-PDGF alpha receptor/PDGFRA Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10556MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human extracellular domain PDGFRa / CD140a (rh PDGFRa; Catalog#10556-H08H; NP_006197.1; Met 1-Glu 524). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1471