Learn More
Sino Biological Anti-PDGF alpha receptor/PDGFRA Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10556MM0720
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PDGFRA / CD140a (rh PDGFRA / CD140a; Catalog#10556-H08H; NP_006197.1; Met1-Glu524). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9074