Learn More
Sino Biological Anti-PDGF alpha receptor/PDGFRA Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10556MM02F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PDGFRA / CD140a (rh PDGFRA / CD140a; Catalog#10556-H08H; NP_006197.1; Met1-Glu524) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1501