Learn More
Sino Biological Human PDGFRA PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10556T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PDGFRA (rh PDGFRA; Catalog#10556-H08H; NP_006197.1; Met1-Glu524). PDGFRA specific IgG was purified by Human PDGFRA affinity chromatography.
Catalog No. 50-237-0824