Learn More
Sino Biological Human PDGFRB MAb 25TS (Sino Biological)
Supplier:  Sino Biological 10514R206A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human PDGFRB/CD140b (rh PDGFRB/CD140b; Catalog#10514-H08H; NP_002600.1; Met1-Lys531) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-237-1440