Learn More
Sino Biological Recombinant Anti-PDGF beta receptor/PDGFRB Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10514R206F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human PDGFRB / CD140b (rh PDGFRB / CD140b; Catalog#10514-H08H; NP_002600.1; Met1-Lys531) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-2436