Learn More
Sino Biological Human PDGFRB MAb 100TS (Sino Biological)
Supplier:  Sino Biological 10890RP02100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human PDGFRB / CD140b (rh PDGFRB / CD140b; Catalog#10514-H08H; NP_002600.1; Met1-Lys531) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-4030