Learn More
Sino Biological Anti-PDGF beta receptor/PDGFRB Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10514RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PDGFRb extracellular domain (rh PDGFRb; Catalog#10514-H08H; NP_002600.1; Met 1-Lys 531). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8336