Learn More
Sino Biological Anti-ERP72 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 310901T36100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human ERP72 / PDIA4 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3779