Learn More
Sino Biological Human PDIA4 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13333T28100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ERP72 / PDIA4 (rh ERP72 / PDIA4; Catalog#13333-H08H; P13667; Met1-Thr641). ERP72 / PDIA4 specific IgG was purified by Human ERP72 / PDIA4 affinity chromatography.
Catalog No. 50-237-3269