Learn More
Sino Biological Human PDPK1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 109876T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PDPK1, and purified by antigen affinity chromatography.
Catalog No. 50-237-2917