Learn More
Sino Biological Anti-Podoplanin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11065T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Podoplanin/PDPN (rh Podoplanin/PDPN; Q86YL7-1; Met1-Lys123). Podoplanin/PDPN specific IgG was purified by Human Podoplanin/PDPN affinity chromatography.
Catalog No. 50-241-0493