Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PDSS1 (aa 35-122) Control Fragment Recombinant Protein

Catalog No. RP104758
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104758 Supplier Invitrogen™ Supplier No. RP104758
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (53%), Rat (53%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65340. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PDSS1 is an enzyme that elongates the prenyl side-chain of coenzyme Q, or ubiquinone, one of the key elements in the respiratory chain. PDSS1 catalyzes the formation of all trans-polyprenyl pyrophosphates from isopentyl diphosphate in the assembly of polyisoprenoid side chains, the first step in coenzyme Q biosynthesis. The protein may be peripherally associated with the inner mitochondrial membrane, though no transit peptide has been definitively identified to date. Defects in PDSS1 gene are a cause of coenzyme Q10 deficiency.The protein encoded by this gene is an enzyme that elongates the prenyl side-chain of coenzyme Q, or ubiquinone, one of the key elements in the respiratory chain. The gene product catalyzes the formation of all trans-polyprenyl pyrophosphates from isopentyl diphosphate in the assembly of polyisoprenoid side chains, the first step in coenzyme Q biosynthesis. The protein may be peripherally associated with the inner mitochondrial membrane, though no transit peptide has been definitively identified to date. Defects in this gene are a cause of coenzyme Q10 deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5T2R2
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 23590
Name Human PDSS1 (aa 35-122) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2610203G20Rik; 2700031G06Rik; All-trans-decaprenyl-diphosphate synthase subunit 1; COenzyme Q (ubiquinone) biosynthesis family member (coq-1)-like; coenzyme Q1 homolog; COQ1; COQ10D2; decaprenyl diphdsphate synthase subunit1; decaprenyl diphosphate synthase subunit 1; decaprenyl pyrophosphate synthase subunit 1; decaprenyl pyrophosphate synthetase subunit 1; decaprenyl-diphosphate synthase subunit 1; DPS; Dps1; hDPS1; hypothetical protein LOC550349; LOW QUALITY PROTEIN: decaprenyl-diphosphate synthase subunit 1; mDLP1; MGC70953; mSPS1; PDSS1; polyprenyl pyrophosphate synthetase; prenyl (decaprenyl) diphosphate synthase, subunit 1; prenyl (solanesyl) diphosphate synthase, subunit 1; prenyl diphosphate synthase, subunit 1; RP13-16H11.3; solanesyl-diphosphate synthase subunit 1; SPS; Sps1; subunit 1 of decaprenyl diphosphate synthase; subunit 1 of solanesyl diphosphate synthase; TPRT; TPT; TPT 1; trans-prenyltransferase; trans-prenyltransferase (TPT); trans-prenyltransferase 1; wu:fc11f12; wu:fd05d05; zgc:112058
Common Name PDSS1
Gene Symbol PDSS1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SAAAEVRAQVHRRKGLDLSQIPYINLVKHLTSACPNVCRISRFHHTTPDSKTHSGEKYTDPFKLGWRDLKGLYEDIRKELLISTSELK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.