Learn More
Sino Biological Anti-PDX1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100419T36100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human PDX1, and purified by antigen affinity chromatography.
Catalog No. 50-241-0513