Learn More
Sino Biological Human PDYN PAb 20ul (Sino Biological)
Supplier:  Sino Biological 80442R02H100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human PDYN/ProDynorphin fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-4183