Learn More
Sino Biological Human PDZD11 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11360T26100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PDZD11 (rh PDZD11; Catalog#12188-H07E; NP_057568.1; Asp2-His140). PDZD11 specific IgG was purified by Human PDZD11 affinity chromatography.
Catalog No. 50-237-4034