Learn More
Sino Biological Human PDZK1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 202029T44100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human PDZK1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-0588