Learn More
Sino Biological Human PEBP1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 14582T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PEBP1 (rh PEBP1; Catalog#14582-HNAE; NP_002558.1; Met1-Lys187). PEBP1 specific IgG was purified by Human PEBP1 affinity chromatography.
Catalog No. 50-237-1596