Learn More
Sino Biological Anti-CD31/PECAM-1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10148MM1220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD31 / PECAM-1 (rh CD31 / PECAM-1; Catalog#10148-H08H; EAW94208.1; Met1-Lys601). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-7965