Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PEN2 Synthetic Peptide

Catalog No. PEP0426 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0426 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0426 Supplier Invitrogen™ Supplier No. PEP0426
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 13 amino acids near the carboxy terminus of human PEN2. PEP-0426 can be used as a blocking peptide with polyclonal antibody PA5-20301.

Presenilins, which are components of the gamma-secretase protein complex, are required for intramembranous processing of some type I transmembrane proteins, such as the Notch proteins and the beta-amyloid precursor protein. Signaling by Notch receptors mediates a wide range of developmental cell fates. Processing of the beta-amyloid precursor protein generates neurotoxic amyloid beta peptides, the major component of senile plaques associated with Alzheimer's disease. This gene encodes a protein that is required for Notch pathway signaling, and for the activity and accumulation of gamma-secretase.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 55851
Species Human
Purification Method purified
Accession Number Q9NZ42
Common Name PEN2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 1700023M09Rik; gamma-secretase subunit PEN-2; hematopoietic stem/progenitor cells protein MDS033; liver regeneration-related protein LRRGT00140; LRRGT00140 mRNA; MDS033; MSTP064; Pen2; Pen-2; presenilin enhancer 2; presenilin enhancer 2 homolog; presenilin enhancer 2 homolog (C. elegans); presenilin enhancer gamma secretase subunit; presenilin enhancer gamma-secretase subunit; presenilin enhancer protein 2; presenilin enhancer, gamma-secretase subunit; Psenen; RGD1312037
Gene Symbol PSENEN
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.