Learn More
Biomatik Corporation Human Peptidase D (PEPD) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12089
Encompass_Preferred
Human Peptidase D (PEPD) Mini sample ELISA Kit for detecting PEPD in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12955. Gene ID: 5184. Target Synonyms: PRD; Xaa-Pro Dipeptidase; Prolidase; Imidodipeptidase; Proline dipeptidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6160