Learn More
Biomatik Corporation Human Peptidase Inhibitor 15 (PI15) ELISA Kit
Supplier:  Biomatik Corporation EKU12319
Encompass_Preferred
Human Peptidase Inhibitor 15 (PI15) ELISA Kit for detecting PI15 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O43692. Gene ID: 51050. Target Synonyms: P25TI; CRISP-8; 25 kDa trypsin inhibitor; Cysteine-rich secretory protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5677