Learn More
Biomatik Corporation Human Peptide YY 3 (PYY3) ELISA Kit
Supplier:  Biomatik Corporation EKU06518
Encompass_Preferred
Human Peptide YY 3 (PYY3) ELISA Kit for detecting PYY3 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 12.35-1000pg/ml. Sensitivity: 4.82pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q5JQD4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4368