Learn More
Biomatik Corporation Human Peptidyl Arginine Deiminase Type III (PADI3) ELISA Kit
Supplier:  Biomatik Corporation EKU08440
Encompass_Preferred
Human Peptidyl Arginine Deiminase Type III (PADI3) ELISA Kit for detecting PADI3 in saliva, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ULW8. Target Synonyms: PAD3; PDI3; Peptidylarginine deiminase III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6415