Learn More
Biomatik Corporation Human Peptidyl Arginine Deiminase Type VI (PADI6) ELISA Kit
Supplier:  Biomatik Corporation EKU06524
Encompass_Preferred
Human Peptidyl Arginine Deiminase Type VI (PADI6) ELISA Kit for detecting PADI6 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6TGC4. Gene ID: 353238. Target Synonyms: Peptidylarginine deiminase VI; Peptidyl arginine deiminase-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4816