Learn More
Biomatik Corporation Human Pericentrin (PCNT) ELISA Kit
Supplier:  Biomatik Corporation EKU09134
Encompass_Preferred
Human Pericentrin (PCNT) ELISA Kit for detecting PCNT in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95613. Gene ID: 5116. Target Synonyms: KEN; PCN; PCNT2; PCNTB; PCTN2; SCKL4; Kendrin; Seckel Syndrome 4; Pericentrin-B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8562