Learn More
Biomatik Corporation Human Perilipin 3 (PLIN3) ELISA Kit
Supplier:  Biomatik Corporation EKU06532
Encompass_Preferred
Human Perilipin 3 (PLIN3) ELISA Kit for detecting PLIN3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60664. Gene ID: 10226. Target Synonyms: M6PRBP1; TIP47; PP17; Mannose-6-Phosphate Receptor Binding Protein 1; Placental Protein 17; 47 kDa mannose 6-phosphate receptor-binding protein; Cargo selection TIP47. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5332