Learn More
Biomatik Corporation Human Period Circadian Protein 1 (PER1) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10957
Encompass_Preferred
Human Period Circadian Protein 1 (PER1) High-sensitive ELISA Kit for detecting PER1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15534. Gene ID: 5187. Target Synonyms: PER; RIGUI; hPER; Circadian clock protein PERIOD 1; Circadian pacemaker protein Rigui. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8262