Learn More
Meso Scale Discovery R-PLEX Human Periostin/OSF-2 Assay, Singleplex, SECTOR plates, 5 plates, compatible with MESO SECTOR instruments, Research Use Only
SDP
Catalog No. NC2747655
Encompass
The MESO SCALE DISCOVERY R-PLEX Human Periostin/OSF-2 Assay is a specialized immunoassay designed for the detection of human Periostin/OSF-2. Utilizing advanced electrochemiluminescence technology, this assay ensures high sensitivity and reliability for biomarker discovery and development. The complete kit includes all essential components for easy assay setup and execution, making it ideal for research applications.
  • Designed for research use only
  • Electrochemiluminescence assay principle
  • Singleplex format for specific analyte detection
  • Compatible with MESO SECTOR instruments
  • Comes with biotinylated capture and SULFO-TAG™ detection antibodies
  • Includes calibrator, diluents, and MSD GOLD™ Read Buffer B
  • Lower limit of detection calculated at 2.5 standard deviations above background
  • Supports multiplexing with other analytes using R-PLEX Assay Designer
  • Quick and easy assay development process
Catalog No. NC2747655 Supplier Meso Scale Discovery Supplier No. K1519WR2
Add to Cart
Add to Cart
The MESO SCALE DISCOVERY R-PLEX Human Periostin/OSF-2 Assay is a specialized immunoassay designed for the detection of human Periostin/OSF-2. Utilizing advanced electrochemiluminescence technology, this assay ensures high sensitivity and reliability for biomarker discovery and development. The complete kit includes all essential components for easy assay setup and execution, making it ideal for research applications.
  • Designed for research use only
  • Electrochemiluminescence assay principle
  • Singleplex format for specific analyte detection
  • Compatible with MESO SECTOR instruments
  • Comes with biotinylated capture and SULFO-TAG™ detection antibodies
  • Includes calibrator, diluents, and MSD GOLD™ Read Buffer B
  • Lower limit of detection calculated at 2.5 standard deviations above background
  • Supports multiplexing with other analytes using R-PLEX Assay Designer
  • Quick and easy assay development process