Learn More
STEMCELL Technologies Human Peripheral Blood B Cells, Frozen, Size: 2 x 10^7 cells
SDP
Catalog No. 501975795
Encompass
Human primary B cells derived from peripheral blood (PB) are isolated from mononuclear cells (MNCs) using the negative immunomagnetic selection technique. Cells are untouched and ready for downstream applications. PB was collected using acid-citrate-dextrose solution A (ACDA) as the anticoagulant.Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/primary-cells-faq) on Primary Cells.
Catalog No. 50-197-5795 Supplier STEMCELL Technologies Supplier No. 70023.1
Human primary B cells derived from peripheral blood (PB) are isolated from mononuclear cells (MNCs) using the negative immunomagnetic selection technique. Cells are untouched and ready for downstream applications. PB was collected using acid-citrate-dextrose solution A (ACDA) as the anticoagulant.Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/primary-cells-faq) on Primary Cells.