Learn More
STEMCELL Technologies Human Peripheral Blood CD4+CD45RO+ T Cells, Frozen, Size: 5 x 10^6 cells
SDP
Supplier:  STEMCELL Technologies 70031
Encompass
Primary human CD4+CD45RO+ T cells were isolated from peripheral blood (PB) mononuclear cells (MNCs) using negative immunomagnetic separation techniques. PB was collected using acid-citrate-dextrose solution A (ACDA) as the anticoagulant. CD4 and CD45RO are co-expressed on memory CD4+ T cells.Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/primary-cells-faq) on Primary Cells.
Catalog No. 50-197-5797