Learn More
STEMCELL Technologies Human Peripheral Blood CD56+ Cells, Frozen, Size: 5 x 10^6 cells
SDP
Supplier:  STEMCELL Technologies 70037
Encompass
Primary human CD56+ cells were isolated from peripheral blood (PB) mononuclear cells (MNCs) using positive immunomagnetic separation techniques. PB was collected using acid-citrate-dextrose solution A (ACDA) as the anticoagulant. CD56 antigen is expressed primarily on natural killer (NK) cells, as well as NKT cells in PB.Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/primary-cells-faq) on Primary Cells.NOTE: This product was formerly named "Human Peripheral Blood CD56+ NK Cells, Frozen"; the product itself and manufacturing procedures have not changed, but the name has been updated to more accurately reflect the cell type provided.
Catalog No. NC1524392
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov