Learn More
STEMCELL Technologies Human Peripheral Blood CD8+ T Cells, Frozen, Size: 1 x 10^7 cells
SDP
Supplier:  STEMCELL Technologies 70027
Encompass
Primary human CD8+ T cells were isolated from peripheral blood (PB) mononuclear cells (MNCs) using negative immunomagnetic separation techniques. PB was collected using the anticoagulant acid-citrate-dextrose solution A (ACDA).Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) on Primary Cells.
Catalog No. NC1239787
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov