Learn More
ORGANABIO LLC Human Peripheral Blood Leukopak / Cryopreserved / Single Donor / Quarter bag (2.5x10E9 TNC) / OrganaBio LEUKOPAC / Research Use Only
Catalog No. NC3997780
Encompass
Research grade leukopaks from recallable / HLA-type healthy donors. Comprehensive characterization CBC / immunophenotyping (CD45 / CD14 / CD19 / CD56 / CD3 / CD3CD4 / CD3CD8) / high-resolution HLA (Class I and II) and infectious disease testing. Available as fresh whole (10x10E9 TNC) / frozen quarter (2.5x10E9 TNC) / and frozen half (5x10E9 TNC) bags. OrganaBio research grade leukopaks are collected from normal / healthy adult donors at FDA-registered apheresis center. Each unit is comprehensively characterized including CBC / immunophenotyping / and infectious disease testing. Detailed donor demographics-including high-resolution HLA typing / age / sex / BMI / race / ethnicity / and blood type-are provided with every unit.
Catalog No. NC3997780 Supplier ORGANABIO LLC Supplier No. LKPPB004
Research grade leukopaks from recallable / HLA-type healthy donors. Comprehensive characterization CBC / immunophenotyping (CD45 / CD14 / CD19 / CD56 / CD3 / CD3CD4 / CD3CD8) / high-resolution HLA (Class I and II) and infectious disease testing. Available as fresh whole (10x10E9 TNC) / frozen quarter (2.5x10E9 TNC) / and frozen half (5x10E9 TNC) bags. OrganaBio research grade leukopaks are collected from normal / healthy adult donors at FDA-registered apheresis center. Each unit is comprehensively characterized including CBC / immunophenotyping / and infectious disease testing. Detailed donor demographics-including high-resolution HLA typing / age / sex / BMI / race / ethnicity / and blood type-are provided with every unit.