Learn More
STEMCELL Technologies Human Peripheral Blood Mononuclear Cells, Frozen
SDP
Catalog No. NC1253358
Encompass
Streamline your assays with ready-to-use, ethically sourced, primary human mononuclear cells. With personalized service, flexible delivery times, and the option to reserve entire lots to prescreen cells for applications, we help you get the cells you need. Isolated from peripheral blood leukapheresis samples using density gradient separation and/or red blood cell lysis and cryopreserved in animal component-free CryoStorCS10 medium (Catalog #07930), cells are collected using Institutional Review Board (IRB)-approved consent forms and protocols. Additional documentation and high-resolution HLA typing (Class I and Class II alleles and CMV status) are available upon request. Acid-citrate-dextrose solution A (ACDA) is added during collection as an anticoagulant. Donor specifications (e.g. BMI category, smoking status, ethnicity, etc.) can be requested. Donors are screened for HIV-1, HIV-2, hepatitis B, and hepatitis C.Certain products are only available in select territories.
This item is not returnable. View return policy
Catalog No. NC1253358 Supplier STEMCELL Technologies Supplier No. 70025.3
Streamline your assays with ready-to-use, ethically sourced, primary human mononuclear cells. With personalized service, flexible delivery times, and the option to reserve entire lots to prescreen cells for applications, we help you get the cells you need. Isolated from peripheral blood leukapheresis samples using density gradient separation and/or red blood cell lysis and cryopreserved in animal component-free CryoStorCS10 medium (Catalog #07930), cells are collected using Institutional Review Board (IRB)-approved consent forms and protocols. Additional documentation and high-resolution HLA typing (Class I and Class II alleles and CMV status) are available upon request. Acid-citrate-dextrose solution A (ACDA) is added during collection as an anticoagulant. Donor specifications (e.g. BMI category, smoking status, ethnicity, etc.) can be requested. Donors are screened for HIV-1, HIV-2, hepatitis B, and hepatitis C.Certain products are only available in select territories.