Learn More
STEMCELL Technologies Human Peripheral Blood NK Cells, Frozen, Size: 5 x 10^6 cells
SDP
Catalog No. NC1169684
Encompass
Primary human NK cells were isolated from peripheral blood (PB) mononuclear cells (MNCs) using negative immunomagnetic separation techniques. PB was collected using acid-citrate-dextrose solution A (ACDA) as the anticoagulant.Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/primary-cells-faq) on Primary Cells.
Catalog No. NC1169684 Supplier STEMCELL Technologies Supplier No. 70036
Primary human NK cells were isolated from peripheral blood (PB) mononuclear cells (MNCs) using negative immunomagnetic separation techniques. PB was collected using acid-citrate-dextrose solution A (ACDA) as the anticoagulant.Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/primary-cells-faq) on Primary Cells.