Learn More
STEMCELL Technologies Human Peripheral Blood Plasma, Frozen, Size: 30 mL
SDP
Supplier:  STEMCELL Technologies 70039.3
Encompass
Primary human plasma has been isolated from peripheral blood (PB) using centrifugation. PB was collected using one of the following anticoagulants: acid-citrate-dextrose (ACD), acid-citrate-dextrose solution A (ACDA), citrate-phosphate-dextrose (CPD), citrate-phosphate-double-dextrose (CP2D), or citrate-phosphate-dextrose-adenine (CPDA).Plasma was obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.
Catalog No. 50-197-5805