Learn More
Biomatik Corporation Human Periplakin (PPL) ELISA Kit
Supplier:  Biomatik Corporation EKU06545
Encompass_Preferred
Human Periplakin (PPL) ELISA Kit for detecting PPL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60437. Gene ID: 5493. Target Synonyms: 190 kDa paraneoplastic pemphigus antigen; 195 kDa cornified envelope precursor protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6606