Learn More
Biomatik Corporation Human Peroxisomal Biogenesis Factor 1 (PEX1) ELISA Kit
Supplier:  Biomatik Corporation EKU12006
Encompass_Preferred
Human Peroxisomal Biogenesis Factor 1 (PEX1) ELISA Kit for detecting PEX1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O43933. Gene ID: 5189. Target Synonyms: ZWS1; Peroxin-1; Peroxisome biogenesis disorder protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6200