Learn More
Biomatik Corporation Human Pescadillo Homolog 1 (PES1) ELISA Kit
Supplier:  Biomatik Corporation EKU06574
Encompass_Preferred
Human Pescadillo Homolog 1 (PES1) ELISA Kit for detecting PES1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00541. Gene ID: 23481. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6217