Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PF4 (P02776, 70 amino acids) native Protein from human platelets

Catalog No. 89956489 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-489 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-956-489 Supplier Abnova™ Supplier No. P4061

Used for Func, SDS-PAGE

Platelet factor-4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair (Eisman et al., 1990 [PubMed 1695112]).[supplied by OMIM]

Specifications

Accession Number P02776
For Use With (Application) Functional Assay, SDS-PAGE
Formulation Lyophilized from PBS, pH 7.4
Gene ID (Entrez) 5196
Molecular Weight (g/mol) 7.8kDa
Name Human PF4 (P02776, 70 amino acids) native Protein from human platelets
Quantity 20 μg
Storage Requirements Store at −20°C on dry atmosphere.After reconstitution with sterilized water, store at −20°C.Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CXCL4, MGC138298, SCYB4
Common Name PF4
Gene Symbol PF4
Biological Activity Binding of HIT-antibodies.
Species Human
Protein Tag Untagged
Expression System Native protein purified from human platelets
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.